Assay Details
Target Gene Details
Entrez Gene ID: | 84650 |
Gene Name: | emopamil binding protein like |
Gene Aliases: |
EBRP |
Location: |
Chr.13:49660676-49691487 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EBPL | NM_001278636.1 | NP_001265565.1 | ||
| NM_032565.4 | NP_115954.1 | |||
| NR_103802.1 | ||||
| NR_103803.1 | ||||
| AF243433.1 | AAK28348.1 | |||
| BC018478.1 | AAH18478.1 | |||
| BC073152.1 | AAH73152.1 | |||
| BC092471.1 | AAH92471.1 | |||
| BY799168.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3580633 | Chr.13:49683591 - 49701621 on Build GRCh38 | Loss |
|
| esv3632050 | Chr.13:49539436 - 49701818 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map