Assay Details
Target Gene Details
Entrez Gene ID: | 23348 |
Gene Name: | dedicator of cytokinesis 9 |
Gene Aliases: |
ZIZ1, ZIZIMIN1 |
Location: |
Chr.13:98793487-99086406 on Build GRCh38 |
Assay Gene Location: | Within Intron 43 |
Target Gene Details
Entrez Gene ID: | 100874096 |
Gene Name: | DOCK9 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:98832084-98834629 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DOCK9-AS1 | NR_046663.1 |
More Information
Set Membership: |
|

Genomic Map