Assay Details
Target Gene Details
Entrez Gene ID: | 57161 |
Gene Name: | pellino E3 ubiquitin protein ligase family member 2 |
Gene Aliases: |
- |
Location: |
Chr.14:56118330-56301586 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PELI2 | NM_021255.2 | NP_067078.1 | ||
| XM_005267890.4 | XP_005267947.1 | |||
| XM_011536990.2 | XP_011535292.1 | |||
| XM_011536992.2 | XP_011535294.1 | |||
| XM_017021479.1 | 1 | 6497 | XP_016876968.1 | |
| AF302502.1 | AAG15390.1 | |||
| AK315727.1 | ||||
| BC009476.2 | AAH09476.1 |
More Information
Set Membership: |
|

Genomic Map