Assay Details
Target Gene Details
Entrez Gene ID: | 3705 |
Gene Name: | inositol-tetrakisphosphate 1-kinase |
Gene Aliases: |
ITRPK1 |
Location: |
Chr.14:92936914-93115918 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITPK1 | AY129028.1 | 1 | 2452 | AAM98771.1 |
| BC003622.2 | 11 | 2298 | AAH03622.1 | |
| BC007428.2 | 11 | 2298 | AAH07428.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892576 | Chr.14:92932504 - 92938764 on Build GRCh38 | Loss |
|
| nsv1400 | Chr.14:92928206 - 92973049 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map