Assay Details
Target Gene Details
Entrez Gene ID: | 100507466 |
Gene Name: | DPH6 antisense RNA 1 (head to head) |
Gene Aliases: |
ATPBD4-AS1 |
Location: |
Chr.15:35546195-35859001 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPH6-AS1 | NR_038251.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs74838585] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map