Assay Details
Target Gene Details
Entrez Gene ID: | 64963 |
Gene Name: | mitochondrial ribosomal protein S11 |
Gene Aliases: |
HCC-2, MRP-S11, S11mt |
Location: |
Chr.15:88467453-88480767 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS11 | NM_001321970.1 | 6 | 2300 | NP_001308899.1 |
| NM_001321972.1 | 6 | 3424 | NP_001308901.1 | |
| NM_022839.4 | 6 | 2303 | NP_073750.2 | |
| NM_176805.3 | 5 | 2204 | NP_789775.1 | |
| NR_135915.1 | 5 | 2284 | ||
| NR_135917.1 | 6 | 2283 | ||
| NR_135918.1 | 6 | 2241 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833085 | Chr.15:88365076 - 88521662 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map