Assay Details
Target Gene Details
Entrez Gene ID: | 6938 |
Gene Name: | transcription factor 12 |
Gene Aliases: |
CRS3, HEB, HTF4, HsT17266, TCF-12, bHLHb20 |
Location: |
Chr.15:56918090-57291261 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3569642 | Chr.15:56740239 - 56941457 on Build GRCh38 | Gain |
|
| nsv833020 | Chr.15:56763749 - 56923870 on Build GRCh38 | Gain+Loss |
|
| nsv457162 | Chr.15:56790808 - 56933568 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map