Assay Details
Target Gene Details
Entrez Gene ID: | 6938 |
Gene Name: | transcription factor 12 |
Gene Aliases: |
CRS3, HEB, HTF4, HsT17266, TCF-12, bHLHb20 |
Location: |
Chr.15:56918090-57291261 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
More Information
Additional Information:
For this assay, SNP(s) [rs115956520] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map