Assay Details
Target Gene Details
Entrez Gene ID: | 9333 |
Gene Name: | transglutaminase 5 |
Gene Aliases: |
PSS2, TGASE5, TGASEX, TGM6, TGMX, TGX |
Location: |
Chr.15:43232595-43266857 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGM5 | NM_004245.3 | NP_004236.1 | ||
| NM_201631.3 | NP_963925.2 | |||
| XM_011522230.2 | XP_011520532.1 | |||
| XM_017022729.1 | XP_016878218.1 | |||
| AF035960.1 | AAC02978.1 | |||
| AF035961.1 | AAC02979.1 | |||
| AK298479.1 | ||||
| BC119009.1 | ||||
| BC122859.1 |
More Information
Set Membership: |
|

Genomic Map