Assay Details
Target Gene Details
Entrez Gene ID: | 4916 |
Gene Name: | neurotrophic receptor tyrosine kinase 3 |
Gene Aliases: |
GP145-TrkC, TRKC, gp145(trkC) |
Location: |
Chr.15:87859749-88256796 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTRK3 | NM_001007156.2 | NP_001007157.1 | ||
| NM_001012338.2 | NP_001012338.1 | |||
| NM_001243101.1 | NP_001230030.1 | |||
| NM_002530.3 | NP_002521.2 | |||
| XM_006720543.3 | 1 | 127 | XP_006720606.1 | |
| XM_006720544.3 | 1 | 127 | XP_006720607.1 | |
| XM_006720545.3 | 1 | 127 | XP_006720608.1 | |
| XM_006720548.3 | 1 | 127 | XP_006720611.1 | |
| XM_006720549.3 | 1 | 127 | XP_006720612.1 | |
| XM_006720550.3 | 1 | 127 | XP_006720613.1 | |
| XM_011521637.2 | 1 | 127 | XP_011519939.1 | |
| XM_011521638.2 | 1 | 127 | XP_011519940.1 | |
| XM_017022240.1 | XP_016877729.1 | |||
| XM_017022242.1 | 1 | 127 | XP_016877731.1 | |
| XM_017022247.1 | 1 | 127 | XP_016877736.1 | |
| XM_017022248.1 | 1 | 127 | XP_016877737.1 | |
| XM_017022249.1 | 1 | 127 | XP_016877738.1 | |
| XM_017022250.1 | 1 | 127 | XP_016877739.1 | |
| XM_017022253.1 | 1 | 127 | XP_016877742.1 | |
| XM_017022254.1 | 1 | 127 | XP_016877743.1 | |
| BI962878.1 | ||||
| CK903553.1 | ||||
| DA789502.1 |
Target Gene Details
Entrez Gene ID: | 283738 |
Gene Name: | NTRK3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:88252730-88271066 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTRK3-AS1 | NR_038229.1 | |||
| AK097486.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952021 | Chr.15:88252070 - 88257269 on Build GRCh38 | Deletion |
|
| nsv827427 | Chr.15:88254355 - 88258505 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map