Assay Details
Target Gene Details
Entrez Gene ID: | 57188 |
Gene Name: | ADAMTS like 3 |
Gene Aliases: |
ADAMTSL-3 |
Location: |
Chr.15:83654086-84039842 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAMTSL3 | NM_001301110.1 | NP_001288039.1 | ||
| NM_207517.2 | NP_997400.2 | |||
| XM_011521822.2 | XP_011520124.1 | |||
| XM_011521823.2 | XP_011520125.1 | |||
| XM_011521824.2 | XP_011520126.1 | |||
| XM_011521825.2 | XP_011520127.1 | |||
| XM_017022434.1 | XP_016877923.1 | |||
| AF237652.1 | AAK15041.1 | |||
| AK092324.1 | ||||
| AK308845.1 | ||||
| BC128389.1 | ||||
| BC128390.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160314 | Chr.15:83632364 - 83706547 on Build GRCh38 | Duplication |
|
| nsv833072 | Chr.15:83588968 - 83746139 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72761824] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map