Assay Details
Target Gene Details
Entrez Gene ID: | 2958 |
Gene Name: | general transcription factor IIA subunit 2 |
Gene Aliases: |
HsT18745, T18745, TF2A2, TFIIA, TFIIA-12, TFIIA-gamma, TFIIAS |
Location: |
Chr.15:59638062-59657541 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTF2A2 | NM_001320929.1 | NP_001307858.1 | ||
| NM_001320930.1 | NP_001307859.1 | |||
| NM_004492.2 | NP_004483.1 | |||
| AK312015.1 | ||||
| BC000287.2 | AAH00287.1 | |||
| BC001919.1 | AAH01919.1 | |||
| CB156109.1 | ||||
| HY002503.1 | ||||
| HY036972.1 | ||||
| U14193.1 | AAA64951.1 | |||
| U21242.1 | AAB58247.1 | |||
| X81713.1 | CAA57357.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527307 | Chr.15:58811129 - 59726288 on Build GRCh38 | Gain |
|
| esv3636647 | Chr.15:59650139 - 59669840 on Build GRCh38 | Gain |
|
| nsv1043693 | Chr.15:59652272 - 59679301 on Build GRCh38 | Loss |
|
| esv3569653 | Chr.15:59636874 - 59670724 on Build GRCh38 | Gain |
|
| nsv569605 | Chr.15:59563027 - 59664354 on Build GRCh38 | Loss |
|
| esv3636646 | Chr.15:59635720 - 59677766 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map