Assay Details
Target Gene Details
Entrez Gene ID: | 8554 |
Gene Name: | protein inhibitor of activated STAT 1 |
Gene Aliases: |
DDXBP1, GBP, GU/RH-II, ZMIZ3 |
Location: |
Chr.15:68054179-68191464 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIAS1 | NM_001320687.1 | NP_001307616.1 | ||
| NM_016166.2 | NP_057250.1 | |||
| XM_011522126.2 | XP_011520428.1 | |||
| XM_011522127.2 | XP_011520429.1 | |||
| XM_017022688.1 | XP_016878177.1 | |||
| XM_017022689.1 | XP_016878178.1 | |||
| XM_017022690.1 | 1 | 382 | XP_016878179.1 | |
| AF077951.1 | AAC36702.1 | |||
| AF167160.1 | AAD49722.1 | |||
| AK094641.1 | ||||
| AK314515.1 | ||||
| AY826819.1 | ||||
| BC118587.1 | ||||
| BC121797.1 | ||||
| EF487997.1 | ||||
| EF487998.1 | ||||
| FJ997900.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951980 | Chr.15:68055363 - 68065062 on Build GRCh38 | Duplication |
|
| nsv1037980 | Chr.15:68061238 - 68145365 on Build GRCh38 | Loss |
|
| nsv517512 | Chr.15:68026786 - 68132927 on Build GRCh38 | Gain |
|
| nsv569803 | Chr.15:67585107 - 68124237 on Build GRCh38 | Gain |
|
| nsv521762 | Chr.15:68057555 - 68179553 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76053662] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map