Assay Details
Target Gene Details
Entrez Gene ID: | 84894 |
Gene Name: | leucine rich repeat and Ig domain containing 1 |
Gene Aliases: |
LERN1, LRRN6A, UNQ201 |
Location: |
Chr.15:77613024-77820900 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINGO1 | NM_001301186.1 | NP_001288115.1 | ||
| NM_001301187.1 | NP_001288116.1 | |||
| NM_001301189.1 | NP_001288118.1 | |||
| NM_001301191.1 | NP_001288120.1 | |||
| NM_001301192.1 | NP_001288121.1 | |||
| NM_001301194.1 | NP_001288123.1 | |||
| NM_001301195.1 | NP_001288124.1 | |||
| NM_001301197.1 | NP_001288126.1 | |||
| NM_001301198.1 | NP_001288127.1 | |||
| NM_001301199.1 | NP_001288128.1 | |||
| NM_001301200.1 | NP_001288129.1 | |||
| XM_011522118.2 | XP_011520420.1 | |||
| XM_017022682.1 | XP_016878171.1 | |||
| BC068558.1 | AAH68558.1 | |||
| BI825536.1 | ||||
| BX355455.2 | ||||
| DA150164.1 | ||||
| DA182630.1 | ||||
| DA183673.1 | ||||
| DA232085.1 | ||||
| DA328735.1 | ||||
| DA410114.1 | ||||
| DA483616.1 | ||||
| DA525603.1 | ||||
| DA787289.1 | ||||
| DB165168.1 |
Target Gene Details
Entrez Gene ID: | 253044 |
Gene Name: | LINGO1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:77641764-77649240 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINGO1-AS1 | NR_045123.1 | 3 | 1023 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833060 | Chr.15:77524312 - 77742862 on Build GRCh38 | Gain |
|
| esv2749898 | Chr.15:75764003 - 77933523 on Build GRCh38 | Deletion |
|
| nsv1040852 | Chr.15:76867896 - 77845199 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map