Assay Details
Target Gene Details
Entrez Gene ID: | 105371031 |
Gene Name: | translation initiation factor IF-2-like |
Gene Aliases: |
- |
Location: |
Chr.15:89399716-89507395 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105371031 |
Target Gene Details
Entrez Gene ID: | 51458 |
Gene Name: | Rh family C glycoprotein |
Gene Aliases: |
C15orf6, PDRC2, RHGK, SLC42A3 |
Location: |
Chr.15:89471407-89496613 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RHCG | NM_001321041.1 | NP_001307970.1 | ||
| NM_016321.2 | NP_057405.1 | |||
| NR_110261.1 | ||||
| AF081497.1 | AAD55748.1 | |||
| AF193809.1 | AAF19372.1 | |||
| AK290899.1 | ||||
| AK298252.1 | ||||
| AK298641.1 | ||||
| AK301640.1 | ||||
| AK313238.1 | ||||
| AY257182.1 | AAP81044.1 | |||
| BC030965.1 | AAH30965.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038140 | Chr.15:89141274 - 89620108 on Build GRCh38 | Gain |
|
| nsv457245 | Chr.15:89464359 - 89517327 on Build GRCh38 | Loss |
|
| esv34025 | Chr.15:89412131 - 89630626 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113679031] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map