Assay Details
Target Gene Details
Entrez Gene ID: | 4205 |
Gene Name: | myocyte enhancer factor 2A |
Gene Aliases: |
ADCAD1, RSRFC4, RSRFC9, mef2 |
Location: |
Chr.15:99565455-99716488 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
More Information
Set Membership: |
|

Genomic Map