Assay Details
Target Gene Details
Entrez Gene ID: | 145781 |
Gene Name: | GRINL1A complex locus 1 |
Gene Aliases: |
GRINL1A, Gcom2, MYZAP, MYZAP-POLR2M, gcom |
Location: |
Chr.15:57591904-57717557 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GCOM1 | NM_001018090.6 | NP_001018100.1 | ||
| NM_001018091.6 | NP_001018101.1 | |||
| NM_001285900.3 | NP_001272829.1 | |||
| NR_104367.2 | ||||
| NR_104368.2 | ||||
| NR_104369.2 | ||||
| NR_104370.2 | ||||
| NR_104371.3 | ||||
| AY207007.1 | AAO39707.1 | |||
| AY208913.1 | AAP41549.1 | |||
| AY331564.1 | AAQ76825.1 | |||
| AY333779.1 | AAQ76826.1 | |||
| AY334560.1 | AAQ76827.1 | |||
| AY334561.1 | AAQ76828.1 | |||
| AY334562.1 | AAQ76829.1 | |||
| AY341345.1 | AAQ76831.1 | |||
| AY353057.1 | AAQ76833.1 | |||
| AY353058.1 | AAQ76834.1 | |||
| AY353060.1 | AAQ76836.1 | |||
| CN284687.1 | ||||
| JF419331.1 |
Target Gene Details
Entrez Gene ID: | 100820829 |
Gene Name: | myocardial zonula adherens protein |
Gene Aliases: |
GCOM1, Gup, MYOZAP |
Location: |
Chr.15:57591904-57685364 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYZAP | NM_001018100.4 | NP_001018110.1 | ||
| NM_152451.7 | NP_689664.3 | |||
| AK055535.1 | BAB70944.1 | |||
| AL833142.1 | ||||
| AY207459.1 | AAP41548.1 | |||
| AY237639.1 | AAP75897.1 | |||
| BC101645.1 | AAI01646.1 | |||
| BC112148.1 | AAI12149.1 | |||
| FJ970029.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045611 | Chr.15:57058767 - 57778609 on Build GRCh38 | Gain |
|
| nsv1054547 | Chr.15:57308290 - 57685461 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs187002071] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map