Assay Details
Target Gene Details
Entrez Gene ID: | 64782 |
Gene Name: | apoptosis enhancing nuclease |
Gene Aliases: |
ISG20L1, pp12744 |
Location: |
Chr.15:88604601-88632281 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AEN | NM_022767.3 | NP_073604.3 | ||
| XM_005254966.1 | XP_005255023.1 | |||
| XM_005254967.1 | XP_005255024.1 | |||
| XM_006720645.3 | XP_006720708.2 | |||
| XM_011521905.2 | XP_011520207.1 | |||
| XM_017022489.1 | XP_016877978.1 | |||
| XM_017022490.1 | XP_016877979.1 | |||
| AF318330.1 | AAL55837.1 | |||
| AK308119.1 | ||||
| BC020988.1 | AAH20988.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs76805089] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map