Assay Details
Target Gene Details
Entrez Gene ID: | 9333 |
Gene Name: | transglutaminase 5 |
Gene Aliases: |
PSS2, TGASE5, TGASEX, TGM6, TGMX, TGX |
Location: |
Chr.15:43232595-43266857 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGM5 | NM_004245.3 | 12 | 2451 | NP_004236.1 |
| NM_201631.3 | 13 | 2697 | NP_963925.2 | |
| XM_011522230.2 | 7 | 1879 | XP_011520532.1 | |
| XM_017022729.1 | 7 | 1903 | XP_016878218.1 | |
| AW467571.1 | 1 | 76 | ||
| CA314704.1 | 1 | 89 |
More Information
Set Membership: |
|

Genomic Map