Assay Details
Target Gene Details
Entrez Gene ID: | 10845 |
Gene Name: | caseinolytic mitochondrial matrix peptidase chaperone subunit |
Gene Aliases: |
- |
Location: |
Chr.15:65148219-65185420 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLPX | NM_006660.4 | 14 | 4578 | NP_006651.2 |
| NR_133680.1 | 14 | 4532 | ||
| AL136922.1 | 12 | 2911 | CAB66856.1 | |
| CA945006.1 | 1 | 211 |
More Information
Set Membership: |
|

Genomic Map