Assay Details
Target Gene Details
Entrez Gene ID: | 663 |
Gene Name: | BCL2 interacting protein 2 |
Gene Aliases: |
BNIP-2, NIP2 |
Location: |
Chr.15:59659146-59689534 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BNIP2 | NM_001320674.1 | NP_001307603.1 | ||
| NM_001320675.1 | NP_001307604.1 | |||
| NM_004330.3 | NP_004321.2 | |||
| AK299628.1 | ||||
| AK308237.1 | ||||
| BC002461.2 | AAH02461.1 | |||
| U15173.1 | AAC00021.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527307 | Chr.15:58811129 - 59726288 on Build GRCh38 | Gain |
|
| nsv1043693 | Chr.15:59652272 - 59679301 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map