Assay Details
Target Gene Details
Entrez Gene ID: | 2558 |
Gene Name: | gamma-aminobutyric acid type A receptor alpha5 subunit |
Gene Aliases: |
- |
Location: |
Chr.15:26866719-26949210 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GABRA5 | NM_000810.3 | NP_000801.1 | ||
| NM_001165037.1 | NP_001158509.1 | |||
| XM_005268258.2 | XP_005268315.1 | |||
| XM_006720459.2 | XP_006720522.1 | |||
| XM_017022055.1 | 10 | 4039 | XP_016877544.1 | |
| A28104.1 | CAA01920.1 | |||
| AK126891.1 | 1 | 1578 | ||
| AK290453.1 | ||||
| AK294303.1 | ||||
| AK303742.1 | ||||
| BC111979.1 | AAI11980.1 | |||
| BC113422.1 | ||||
| BT009830.1 | AAP88832.1 | |||
| L08485.1 | AAA58490.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2668795 | Chr.15:26828644 - 27745218 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map