Assay Details
Target Gene Details
Entrez Gene ID: | 79094 |
Gene Name: | ChaC glutathione specific gamma-glutamylcyclotransferase 1 |
Gene Aliases: |
- |
Location: |
Chr.15:40942135-40956519 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHAC1 | XM_017022563.1 | 1 | 1798 | XP_016878052.1 |
| XM_017022564.1 | XP_016878053.1 | |||
| XM_017022565.1 | 1 | 1798 | XP_016878054.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1504 | Chr.15:40936351 - 40969374 on Build GRCh38 | Insertion |
|
| esv2506979 | Chr.15:40943121 - 40944102 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73402522] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map