Assay Details
Target Gene Details
Entrez Gene ID: | 4916 |
Gene Name: | neurotrophic receptor tyrosine kinase 3 |
Gene Aliases: |
GP145-TrkC, TRKC, gp145(trkC) |
Location: |
Chr.15:87859749-88256796 on Build GRCh38 |
Assay Gene Location: | Within Intron 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTRK3 | NM_001012338.2 | NP_001012338.1 | ||
| NM_001243101.1 | NP_001230030.1 | |||
| NM_002530.3 | NP_002521.2 | |||
| XM_006720543.3 | XP_006720606.1 | |||
| XM_006720544.3 | XP_006720607.1 | |||
| XM_017022240.1 | XP_016877729.1 | |||
| XM_017022241.1 | XP_016877730.1 | |||
| XM_017022243.1 | XP_016877732.1 | |||
| XM_017022244.1 | XP_016877733.1 | |||
| XM_017022245.1 | XP_016877734.1 | |||
| XM_017022246.1 | XP_016877735.1 | |||
| XM_017022251.1 | XP_016877740.1 | |||
| XM_017022252.1 | XP_016877741.1 | |||
| XM_017022253.1 | XP_016877742.1 | |||
| XM_017022254.1 | XP_016877743.1 | |||
| AF052184.1 | ||||
| AK297160.1 | ||||
| AK313646.1 | ||||
| KJ534904.1 | ||||
| KJ534905.1 | ||||
| KJ534906.1 | ||||
| KJ535052.1 | ||||
| S76475.1 | AAB33111.1 | |||
| U05012.1 | AAA75374.1 |
More Information
Set Membership: |
|

Genomic Map