Assay Details
Target Gene Details
Entrez Gene ID: | 101929743 |
Gene Name: | long intergenic non-protein coding RNA 1586 |
Gene Aliases: |
- |
Location: |
Chr.15:88585571-88605110 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC01586 | NR_120369.1 | 3 | 594 | |
| NR_120370.1 | 3 | 584 | ||
| AK054749.1 | 3 | 594 | ||
| BX488539.1 | 3 | 588 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509581 | Chr.15:88514213 - 88597724 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116463403] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map