Assay Details
Target Gene Details
Entrez Gene ID: | 283738 |
Gene Name: | NTRK3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:88252730-88271066 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTRK3-AS1 | NR_038229.1 | |||
| AK097486.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952021 | Chr.15:88252070 - 88257269 on Build GRCh38 | Deletion |
|
| nsv827427 | Chr.15:88254355 - 88258505 on Build GRCh38 | Gain |
|
| nsv1112675 | Chr.15:88256769 - 88257269 on Build GRCh38 | Deletion |
|

Genomic Map