Assay Details
Target Gene Details
Entrez Gene ID: | 283738 |
Gene Name: | NTRK3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:88252730-88271066 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTRK3-AS1 | NR_038229.1 | |||
| AK097486.1 |

Genomic Map