Assay Details
Target Gene Details
Entrez Gene ID: | 9488 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class B |
Gene Aliases: |
GPI-MT-III, PIG-B |
Location: |
Chr.15:55318935-55355648 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGB | NM_004855.4 | NP_004846.4 | ||
| XM_005254795.4 | XP_005254852.1 | |||
| XM_011522235.2 | XP_011520537.1 | |||
| XM_011522236.2 | XP_011520538.1 | |||
| XM_011522237.2 | XP_011520539.1 | |||
| XM_017022730.1 | XP_016878219.1 | |||
| XM_017022731.1 | XP_016878220.1 | |||
| XM_017022732.1 | XP_016878221.1 | |||
| AK223330.1 | BAD97050.1 | |||
| AK293666.1 | ||||
| AK314706.1 | ||||
| BC017711.1 | AAH17711.1 | |||
| D42138.1 | BAA07709.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761899 | Chr.15:53592144 - 55643724 on Build GRCh38 | Gain+Loss |
|
| nsv1043789 | Chr.15:55108674 - 55800256 on Build GRCh38 | Gain |
|
| nsv569523 | Chr.15:55263404 - 55544506 on Build GRCh38 | Gain |
|
| esv3569598 | Chr.15:55270429 - 55410270 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map