Assay Details
Target Gene Details
Entrez Gene ID: | 5604 |
Gene Name: | mitogen-activated protein kinase kinase 1 |
Gene Aliases: |
CFC3, MAPKK1, MEK1, MKK1, PRKMK1 |
Location: |
Chr.15:66386873-66491544 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP2K1 | NM_002755.3 | NP_002746.1 | ||
| XM_011521783.2 | XP_011520085.1 | |||
| XM_017022411.1 | XP_016877900.1 | |||
| XM_017022412.1 | XP_016877901.1 | |||
| AK291500.1 | ||||
| AK294322.1 | ||||
| AK312356.1 | ||||
| BC137459.1 | ||||
| BC139729.1 | ||||
| L05624.1 | AAA36318.1 | |||
| L11284.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833045 | Chr.15:66295248 - 66460419 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs118117574] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map