Assay Details
Target Gene Details
Entrez Gene ID: | 3480 |
Gene Name: | insulin like growth factor 1 receptor |
Gene Aliases: |
CD221, IGFIR, IGFR, JTK13 |
Location: |
Chr.15:98648539-98964530 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGF1R | NM_000875.4 | NP_000866.1 | ||
| NM_001291858.1 | NP_001278787.1 | |||
| AB425196.1 | ||||
| BC113610.1 | ||||
| BC113612.1 | ||||
| BC143721.1 | ||||
| DR007209.1 | ||||
| EU826611.1 | ||||
| EU826612.1 | ||||
| X04434.1 | CAA28030.1 |
Target Gene Details
Entrez Gene ID: | 104472848 |
Gene Name: | IGF1R antisense imprinted non-protein coding RNA |
Gene Aliases: |
IGF1R-AS |
Location: |
Chr.15:98645863-98651221 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IRAIN | NR_126453.1 | 1 | 1290 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv27037 | Chr.15:98646891 - 98650587 on Build GRCh38 | Loss |
|
| dgv4766n54 | Chr.15:98648030 - 98650363 on Build GRCh38 | Loss |
|
| nsv827457 | Chr.15:98645110 - 98651725 on Build GRCh38 | Gain |
|
| dgv4764n54 | Chr.15:98647653 - 98650539 on Build GRCh38 | Gain+Loss |
|
| nsv570701 | Chr.15:98647822 - 98652883 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map