Assay Details
Target Gene Details
Entrez Gene ID: | 4205 |
Gene Name: | myocyte enhancer factor 2A |
Gene Aliases: |
ADCAD1, RSRFC4, RSRFC9, mef2 |
Location: |
Chr.15:99565455-99716488 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MEF2A | XM_005254915.2 | 1 | 54 | XP_005254972.1 |
| XM_011521572.2 | XP_011519874.1 | |||
| XM_011521579.2 | XP_011519881.1 | |||
| XM_011521581.2 | 1 | 54 | XP_011519883.1 | |
| XM_011521582.2 | 1 | 42 | XP_011519884.1 | |
| XM_011521585.2 | 1 | 54 | XP_011519887.1 | |
| XM_011521586.2 | 1 | 54 | XP_011519888.1 | |
| XM_011521587.2 | 1 | 54 | XP_011519889.1 | |
| XM_017022193.1 | XP_016877682.1 | |||
| XM_017022196.1 | 1 | 54 | XP_016877685.1 | |
| XM_017022198.1 | 1 | 54 | XP_016877687.1 | |
| XM_017022200.1 | 1 | 54 | XP_016877689.1 | |
| XM_017022202.1 | 1 | 54 | XP_016877691.1 | |
| XM_017022203.1 | 1 | 54 | XP_016877692.1 | |
| XM_017022204.1 | 1 | 54 | XP_016877693.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1051560 | Chr.15:99296897 - 99584278 on Build GRCh38 | Gain |
|
| esv3892741 | Chr.15:98979139 - 99618239 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map