Assay Details
Target Gene Details
Entrez Gene ID: | 5607 |
Gene Name: | mitogen-activated protein kinase kinase 5 |
Gene Aliases: |
HsT17454, MAPKK5, MEK5, PRKMK5 |
Location: |
Chr.15:67542606-67807117 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP2K5 | NM_001206804.1 | NP_001193733.1 | ||
| NM_002757.3 | NP_002748.1 | |||
| NM_145160.2 | NP_660143.1 | |||
| XM_011521787.2 | XP_011520089.1 | |||
| XM_011521788.2 | XP_011520090.1 | |||
| XM_017022414.1 | XP_016877903.1 | |||
| AK293459.1 | ||||
| AK315435.1 | ||||
| BC008838.2 | AAH08838.1 | |||
| BT006780.1 | AAP35426.1 | |||
| CR542229.1 | CAG47025.1 | |||
| U71087.1 | AAB16851.1 | |||
| U71088.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv569799 | Chr.15:67511204 - 67585107 on Build GRCh38 | Loss |
|
| esv2749801 | Chr.15:67575961 - 67577166 on Build GRCh38 | Deletion |
|
| esv2671430 | Chr.15:67576000 - 67577022 on Build GRCh38 | Deletion |
|
| nsv827374 | Chr.15:67576010 - 67577066 on Build GRCh38 | Loss |
|
| esv27062 | Chr.15:67576254 - 67576890 on Build GRCh38 | Loss |
|
| nsv569800 | Chr.15:67570774 - 67585107 on Build GRCh38 | Loss |
|
| nsv569802 | Chr.15:67576449 - 67585107 on Build GRCh38 | Loss |
|
| esv3552599 | Chr.15:67575957 - 67577138 on Build GRCh38 | Deletion |
|
| esv3636790 | Chr.15:67575945 - 67577058 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map