Assay Details
Target Gene Details
Entrez Gene ID: | 105371031 |
Gene Name: | translation initiation factor IF-2-like |
Gene Aliases: |
- |
Location: |
Chr.15:89399716-89507395 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105371031 | XM_017022800.1 | XP_016878289.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038140 | Chr.15:89141274 - 89620108 on Build GRCh38 | Gain |
|
| nsv516532 | Chr.15:89389564 - 89405705 on Build GRCh38 | Loss |
|
| nsv833086 | Chr.15:89275748 - 89437378 on Build GRCh38 | Loss |
|

Genomic Map