Assay Details
Target Gene Details
Entrez Gene ID: | 100287616 |
Gene Name: | LOXL1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:73917468-73928248 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOXL1-AS1 | NR_040066.1 | |||
| NR_040067.1 | ||||
| NR_040068.1 | ||||
| NR_040069.1 | ||||
| NR_040070.1 | ||||
| AK056376.1 | ||||
| AK056885.1 | ||||
| BX355383.2 | 2 | 446 | ||
| BX366015.2 | ||||
| DA021396.1 | ||||
| DA038997.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1604 | Chr.15:73903279 - 73936860 on Build GRCh38 | Insertion |
|
| nsv1603 | Chr.15:73898881 - 73943392 on Build GRCh38 | Deletion |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|

Genomic Map