Assay Details
Target Gene Details
Entrez Gene ID: | 56924 |
Gene Name: | p21 (RAC1) activated kinase 6 |
Gene Aliases: |
PAK5 |
Location: |
Chr.15:40239091-40277487 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAK6 | NM_001276717.1 | NP_001263646.1 | ||
| NM_001276718.1 | NP_001263647.1 | |||
| NM_020168.5 | NP_064553.1 | |||
| AF276893.1 | AAF82800.1 | |||
| AJ236915.1 | CAC18720.1 | |||
| AK131522.1 | ||||
| BC035596.1 | AAH35596.1 | |||
| DA191028.1 | ||||
| DA229537.1 | ||||
| HY128655.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471240 | Chr.15:40235954 - 40303426 on Build GRCh38 | Loss |
|
| esv34145 | Chr.15:40257416 - 40597626 on Build GRCh38 | Loss |
|
| nsv952611 | Chr.15:40237800 - 40284699 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map