Assay Details
Target Gene Details
Entrez Gene ID: | 64782 |
Gene Name: | apoptosis enhancing nuclease |
Gene Aliases: |
ISG20L1, pp12744 |
Location: |
Chr.15:88604601-88632281 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AEN | XM_011521905.2 | XP_011520207.1 | ||
| XM_017022490.1 | XP_016877979.1 |
Target Gene Details
Entrez Gene ID: | 101929743 |
Gene Name: | long intergenic non-protein coding RNA 1586 |
Gene Aliases: |
- |
Location: |
Chr.15:88585571-88605110 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC01586 | NR_120370.1 | |||
| BX488539.1 |
More Information
Set Membership: |
|

Genomic Map