Assay Details
Target Gene Details
Entrez Gene ID: | 55784 |
Gene Name: | multiple C2 and transmembrane domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.15:94231184-94483952 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCTP2 | XM_005254955.3 | XP_005255012.1 | ||
| XM_006720603.2 | XP_006720666.1 | |||
| XM_011521771.2 | XP_011520073.1 | |||
| XM_011521772.2 | XP_011520074.1 | |||
| XM_017022404.1 | XP_016877893.1 | |||
| XM_017022405.1 | XP_016877894.1 | |||
| AK090622.1 | ||||
| AK300325.1 | ||||
| AL832048.1 | ||||
| AL832717.1 | ||||
| BC041387.1 | AAH41387.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833097 | Chr.15:94162999 - 94351067 on Build GRCh38 | Gain |
|
| nsv1052465 | Chr.15:94011482 - 94804200 on Build GRCh38 | Gain |
|
| dgv759e212 | Chr.15:94236123 - 94285130 on Build GRCh38 | Loss |
|
| nsv570532 | Chr.15:94254313 - 94285237 on Build GRCh38 | Loss |
|
| dgv4730n54 | Chr.15:94237012 - 94285237 on Build GRCh38 | Loss |
|
| nsv1047585 | Chr.15:93942818 - 94435921 on Build GRCh38 | Gain |
|
| nsv516378 | Chr.15:94252429 - 94257220 on Build GRCh38 | Loss |
|
| dgv2665n100 | Chr.15:94233701 - 94285021 on Build GRCh38 | Loss |
|
| esv2761821 | Chr.15:94235856 - 94285033 on Build GRCh38 | Loss |
|
| esv3637308 | Chr.15:94251264 - 94258348 on Build GRCh38 | Gain |
|
| esv3637305 | Chr.15:94229922 - 94285926 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map