Assay Details
Target Gene Details
Entrez Gene ID: | 51621 |
Gene Name: | Kruppel like factor 13 |
Gene Aliases: |
BTEB3, FKLF2, NSLP1, RFLAT-1, RFLAT1 |
Location: |
Chr.15:31326855-31435665 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLF13 | NM_001302461.1 | NP_001289390.1 | ||
| NM_015995.3 | NP_057079.2 | |||
| AF132599.1 | AAD26864.1 | |||
| AF150628.1 | AAD34020.1 | |||
| AI084187.1 | ||||
| BC010438.1 | AAH10438.1 | |||
| BC012741.2 | AAH12741.1 | |||
| BC013946.2 | AAH13946.1 | |||
| KJ534876.1 | ||||
| KJ535015.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3636052 | Chr.15:31325578 - 31378949 on Build GRCh38 | Gain |
|
| nsv516681 | Chr.15:31324471 - 31383250 on Build GRCh38 | Loss |
|
| nsv456748 | Chr.15:31329118 - 31405439 on Build GRCh38 | Loss |
|
| esv34100 | Chr.15:31204311 - 31500388 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map