Assay Details
Target Gene Details
Entrez Gene ID: | 83640 |
Gene Name: | family with sequence similarity 103 member A1 |
Gene Aliases: |
C15orf18, HsT19360, RAM |
Location: |
Chr.15:82986203-82991057 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM103A1 | NM_031452.3 | NP_113640.1 | ||
| AK027095.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3637065 | Chr.15:82942994 - 83064493 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map