Assay Details
Target Gene Details
Entrez Gene ID: | 3480 |
Gene Name: | insulin like growth factor 1 receptor |
Gene Aliases: |
CD221, IGFIR, IGFR, JTK13 |
Location: |
Chr.15:98648539-98964530 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGF1R | NM_000875.4 | NP_000866.1 | ||
| NM_001291858.1 | NP_001278787.1 | |||
| XM_017022139.1 | XP_016877628.1 | |||
| AB425196.1 | ||||
| BC113610.1 | ||||
| BC113612.1 | ||||
| BC143721.1 | ||||
| DR007209.1 | ||||
| EU826611.1 | ||||
| EU826612.1 | ||||
| X04434.1 | CAA28030.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv570701 | Chr.15:98647822 - 98652883 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map