Assay Details
Target Gene Details
Entrez Gene ID: | 4916 |
Gene Name: | neurotrophic receptor tyrosine kinase 3 |
Gene Aliases: |
GP145-TrkC, TRKC, gp145(trkC) |
Location: |
Chr.15:87859749-88256796 on Build GRCh38 |
Assay Gene Location: | Within Exon 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTRK3 | XM_006720543.3 | 17 | 17228 | XP_006720606.1 |
| XM_006720544.3 | 16 | 17204 | XP_006720607.1 | |
| XM_017022240.1 | 18 | 17099 | XP_016877729.1 | |
| XM_017022241.1 | 17 | 16584 | XP_016877730.1 | |
| XM_017022243.1 | 17 | 16580 | XP_016877732.1 | |
| XM_017022244.1 | 17 | 16482 | XP_016877733.1 | |
| XM_017022245.1 | 18 | 16537 | XP_016877734.1 | |
| XM_017022246.1 | 17 | 16638 | XP_016877735.1 | |
| XM_017022251.1 | 10 | 15599 | XP_016877740.1 | |
| XM_017022252.1 | 9 | 15575 | XP_016877741.1 | |
| XM_017022253.1 | 18 | 17270 | XP_016877742.1 | |
| XM_017022254.1 | 18 | 17303 | XP_016877743.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1652 | Chr.15:87829569 - 87863160 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map