Assay Details
Target Gene Details
Entrez Gene ID: | 4756 |
Gene Name: | neogenin 1 |
Gene Aliases: |
IGDCC2, NGN, NTN1R2 |
Location: |
Chr.15:73051715-73305206 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517966 | Chr.15:73057292 - 73196412 on Build GRCh38 | Gain |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map