Assay Details
Target Gene Details
Entrez Gene ID: | 2567 |
Gene Name: | gamma-aminobutyric acid type A receptor gamma3 subunit |
Gene Aliases: |
- |
Location: |
Chr.15:26971282-27537112 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GABRG3 | NM_001270873.1 | NP_001257802.1 | ||
| NM_033223.4 | NP_150092.2 | |||
| XM_011521430.2 | XP_011519732.1 | |||
| BX282033.1 | ||||
| DN990720.1 | ||||
| S82769.1 | AAB39369.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2668795 | Chr.15:26828644 - 27745218 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs141945159] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map