Assay Details
Target Gene Details
Entrez Gene ID: | 283761 |
Gene Name: | long intergenic non-protein coding RNA 928 |
Gene Aliases: |
- |
Location: |
Chr.15:89504930-89524034 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00928 | NR_027074.1 | |||
| NR_027075.1 | ||||
| NR_027076.1 | ||||
| NR_027077.1 | ||||
| AK058110.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv570386 | Chr.15:89464359 - 89517327 on Build GRCh38 | Loss |
|
| nsv1038140 | Chr.15:89141274 - 89620108 on Build GRCh38 | Gain |
|
| nsv1047384 | Chr.15:89498372 - 89571078 on Build GRCh38 | Loss |
|
| esv34025 | Chr.15:89412131 - 89630626 on Build GRCh38 | Loss |
|

Genomic Map