Assay Details
Target Gene Details
Entrez Gene ID: | 145748 |
Gene Name: | LysM domain containing 4 |
Gene Aliases: |
- |
Location: |
Chr.15:99715033-99733561 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LYSMD4 | XM_006720397.3 | XP_006720460.2 | ||
| XM_011521242.2 | XP_011519544.1 | |||
| XM_011521243.2 | XP_011519545.1 | |||
| XM_011521245.2 | XP_011519547.1 | |||
| AK056718.1 | ||||
| BX640928.1 | CAE45966.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4769n54 | Chr.15:99700705 - 100224933 on Build GRCh38 | Gain |
|
| nsv428313 | Chr.15:99695998 - 100216834 on Build GRCh38 | Gain |
|
| esv3637430 | Chr.15:99708759 - 100311912 on Build GRCh38 | Gain |
|
| nsv1045838 | Chr.15:99702862 - 100492013 on Build GRCh38 | Gain |
|

Genomic Map