Assay Details
Target Gene Details
Entrez Gene ID: | 80205 |
Gene Name: | chromodomain helicase DNA binding protein 9 |
Gene Aliases: |
AD013, CHD-9, CReMM, KISH2, PRIC320 |
Location: |
Chr.16:53054991-53327502 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHD9 | NM_025134.4 | NP_079410.4 | ||
| XM_005256168.3 | XP_005256225.1 | |||
| XM_017023719.1 | XP_016879208.1 | |||
| XM_017023720.1 | XP_016879209.1 | |||
| AY647157.1 | AAT66509.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066719 | Chr.16:52261069 - 53107993 on Build GRCh38 | Loss |
|
| nsv833232 | Chr.16:52974938 - 53157922 on Build GRCh38 | Loss |
|
| nsv833233 | Chr.16:53046019 - 53257336 on Build GRCh38 | Gain |
|
| esv3638643 | Chr.16:53057939 - 53061216 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map