Assay Details
Target Gene Details
Entrez Gene ID: | 9100 |
Gene Name: | ubiquitin specific peptidase 10 |
Gene Aliases: |
UBPO |
Location: |
Chr.16:84699949-84779922 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP10 | NM_001272075.1 | NP_001259004.1 | ||
| NM_005153.2 | NP_005144.2 | |||
| NR_073577.1 | ||||
| NR_073578.1 | ||||
| XM_006721332.1 | XP_006721395.1 | |||
| XM_011523440.1 | XP_011521742.1 | |||
| XM_011523441.1 | XP_011521743.1 | |||
| XM_011523443.1 | XP_011521745.1 | |||
| XM_017023863.1 | XP_016879352.1 | |||
| XM_017023864.1 | XP_016879353.1 | |||
| XM_017023865.1 | XP_016879354.1 | |||
| XM_017023867.1 | XP_016879356.1 | |||
| XM_017023868.1 | XP_016879357.1 | |||
| XM_017023869.1 | XP_016879358.1 | |||
| AK293304.1 | ||||
| AK299618.1 | ||||
| AK315570.1 | ||||
| AL162049.1 | CAB82392.2 | |||
| BC000263.1 | AAH00263.1 | |||
| BX537402.1 | CAD97644.1 | |||
| CR749515.1 | CAH18330.1 | |||
| D80012.1 | ||||
| DB077919.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483044 | Chr.16:84166395 - 87066394 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map