Assay Details
Target Gene Details
Entrez Gene ID: | 26156 |
Gene Name: | ribosomal L1 domain containing 1 |
Gene Aliases: |
CSIG, L12, PBK1, UTP30 |
Location: |
Chr.16:11834196-11851585 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RSL1D1 | NM_015659.2 | 9 | 4770 | NP_056474.2 |
| AF074987.1 | 1 | 284 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3637942 | Chr.16:11832692 - 11837191 on Build GRCh38 | Gain |
|
| nsv1043909 | Chr.16:11746054 - 11913578 on Build GRCh38 | Gain |
|
| nsv1120318 | Chr.16:11824982 - 11855178 on Build GRCh38 | Tandem Duplication |
|
| nsv571458 | Chr.16:11831151 - 11993425 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map