Assay Details
Target Gene Details
Entrez Gene ID: | 79091 |
Gene Name: | methyltransferase like 22 |
Gene Aliases: |
C16orf68 |
Location: |
Chr.16:8621640-8673565 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| METTL22 | NM_024109.3 | NP_077014.3 | ||
| NR_134608.1 | ||||
| XM_005255570.3 | XP_005255627.1 | |||
| XM_005255571.2 | XP_005255628.1 | |||
| XM_011522657.2 | XP_011520959.1 | |||
| XM_017023659.1 | XP_016879148.1 | |||
| XM_017023660.1 | XP_016879149.1 | |||
| XM_017023661.1 | XP_016879150.1 | |||
| XM_017023662.1 | XP_016879151.1 | |||
| AK022495.1 | BAB14058.1 | |||
| AK315383.1 | ||||
| BC001908.1 | AAH01908.1 | |||
| DB032220.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047172 | Chr.16:8528343 - 8810411 on Build GRCh38 | Gain |
|
| nsv827546 | Chr.16:8622418 - 8623143 on Build GRCh38 | Gain |
|
| nsv571406 | Chr.16:8596737 - 8690609 on Build GRCh38 | Gain |
|
| nsv1037325 | Chr.16:8615311 - 8688379 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map