Assay Details
Target Gene Details
Entrez Gene ID: | 400506 |
Gene Name: | lysine rich nucleolar protein 1 |
Gene Aliases: |
101F10.1, C16orf88, FAM191A, TSG118 |
Location: |
Chr.16:19706347-19718240 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KNOP1 | XM_005255317.4 | XP_005255374.1 | ||
| XM_006721046.3 | XP_006721109.1 | |||
| XM_011545855.1 | XP_011544157.1 | |||
| XM_017023231.1 | XP_016878720.1 | |||
| XM_017023232.1 | XP_016878721.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv571582 | Chr.16:19038056 - 19790801 on Build GRCh38 | Gain |
|
| nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map